Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027629.3 | NP_081905.1 | 13 - 14 | 2076 | 107 | |
GenBank mRNA | AK016437.1 | - | 11 - 12 | 1435 | 107 | |
AK039189.1 | - | 13 - 14 | 2060 | 107 | ||
AK040513.1 | - | 12 - 13 | 1806 | 107 | ||
BC020351.1 | - | 13 - 14 | 2289 | 107 | ||
BC032150.1 | - | 9 - 10 | 1282 | 5004308 | 107 | |
BC076571.1 | - | 13 - 14 | 1973 | 30356163 | 107 | |
BG071667.2 | - | 1 - 2 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |