Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290789.1 | NP_001277718.1 | 2 - 3 | 1112 | 65 | |
NM_027917.3 | NP_082193.2 | 2 - 3 | 1112 | 65 | ||
GenBank mRNA | AK004934.1 | - | 2 - 3 | 1039 | 65 | |
AK035462.1 | - | 2 - 3 | 1073 | 65 | ||
AK085693.1 | - | 2 - 3 | 1073 | 65 | ||
AK134332.1 | - | 2 - 3 | 1040 | 65 | ||
BC005761.1 | - | 2 - 3 | 1087 | 3601742 | 65 | |
BC031598.1 | - | 2 - 3 | 1004 | 3497170 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |