Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163734.1 | NP_001157206.1 | 6 - 7 | 867 | 71 | |
NM_028244.2 | NP_082520.2 | 9 - 10 | 1183 | 71 | ||
GenBank mRNA | AK011155.1 | - | 2 - 3 | 109 | 71 | |
AK129078.1 | - | 6 - 7 | 867 | 71 | ||
BC016569.1 | - | 9 - 10 | 1183 | 3157173 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |