Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028799.2 | NP_083075.1 | 1 - 2 | 99 | 76 | |
GenBank mRNA | AK009196.2 | - | 1 - 2 | 99 | 76 | |
AK037074.1 | - | 1 - 2 | 153 | 76 | ||
AK079431.1 | - | 1 - 2 | 163 | 76 | ||
AK132482.1 | - | 1 - 2 | 182 | 76 | ||
BC116266.1 | - | 1 - 2 | 82 | 40094000 | 76 | |
BC116267.1 | - | 1 - 2 | 82 | 40094002 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |