Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168321.1 | NP_001161793.1 | 12 - 13 | 4779 | 62 | |
NM_029510.3 | NP_083786.2 | 13 - 14 | 5604 | 62 | ||
NM_175044.3 | NP_778209.2 | 12 - 13 | 5550 | 62 | ||
NM_175045.3 | NP_778210.2 | 13 - 14 | 5502 | 62 | ||
NM_175046.3 | NP_778211.2 | 12 - 13 | 5448 | 62 | ||
GenBank mRNA | AK018034.1 | - | 3 - 4 | 370 | 62 | |
AK031282.1 | - | 5 - 6 | 906 | 62 | ||
AK129398.1 | - | 12 - 13 | 4779 | 62 | ||
AY161170.1 | - | 13 - 14 | 4919 | 62 | ||
AY161171.1 | - | 12 - 13 | 4865 | 62 | ||
AY161172.1 | - | 13 - 14 | 4817 | 62 | ||
AY161173.1 | - | 12 - 13 | 4763 | 62 | ||
BC038637.1 | - | 5 - 6 | 809 | 62 | ||
BC058656.1 | - | 13 - 14 | 5003 | 6412868 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |