Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252437.1 | NP_001239366.1 | 7 - 8 | 967 | 61 | |
NM_029657.4 | NP_083933.1 | 7 - 8 | 964 | 61 | ||
GenBank mRNA | AK011747.2 | - | 7 - 8 | 757 | 61 | |
AK088533.1 | - | 7 - 8 | 747 | 61 | ||
AK129161.1 | - | 4 - 5 | 375 | 61 | ||
AK150176.1 | - | 8 - 9 | 800 | 61 | ||
AK153407.1 | - | 7 - 8 | 788 | 61 | ||
BC046830.1 | - | 7 - 8 | 813 | 6811606 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |