Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289659.1 | NP_001276588.1 | 3 - 4 | 413 | 102 | |
NM_001289660.1 | NP_001276589.1 | 3 - 4 | 380 | 102 | ||
NM_029692.3 | NP_083968.1 | 2 - 3 | 359 | 102 | ||
GenBank mRNA | AK044005.1 | - | 3 - 4 | 328 | 102 | |
AK149444.1 | - | 2 - 3 | 357 | 102 | ||
AY152393.1 | - | 2 - 3 | 358 | 102 | ||
BC027189.1 | - | 3 - 4 | 655 | 4196835 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |