Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172261.3 | NP_758465.2 | 5 - 6 | 2197 | 65 | |
GenBank mRNA | AY508450.1 | - | 5 - 6 | 2197 | 65 | |
BC029672.1 | - | 4 - 5 | 968 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |