Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172262.3 | NP_758466.1 | 11 - 12 | 1425 | 67 | |
GenBank mRNA | AK028553.1 | - | 3 - 4 | 391 | 67 | |
BC023917.1 | - | 11 - 12 | 1366 | 5323227 | 67 | |
BY714054.1 | - | 3 - 4 | 391 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |