Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172287.2 | NP_758491.1 | 4 - 5 | 786 | 92 | |
GenBank mRNA | BC026502.1 | - | 3 - 4 | 614 | 4506974 | 92 |
BC049152.1 | - | 4 - 5 | 786 | 6311485 | 92 | |
BC054072.1 | - | 4 - 5 | 786 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |