Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136471.1 | NP_001129943.1 | 3 - 4 | 253 | 106 | |
NM_172297.1 | NP_758501.2 | 2 - 3 | 270 | 106 | ||
GenBank mRNA | AK134892.1 | - | 3 - 4 | 239 | 106 | |
BC021915.1 | - | 2 - 3 | 270 | 4527178 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |