Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163755.1 | NP_001157227.1 | 4 - 5 | 2523 | 64 | |
NM_001163757.1 | NP_001157229.1 | 4 - 5 | 2523 | 64 | ||
NM_001163758.1 | NP_001157230.1 | 4 - 5 | 2504 | 64 | ||
NM_172446.3 | NP_766034.2 | 4 - 5 | 2576 | 64 | ||
GenBank mRNA | AB185113.1 | - | 4 - 5 | 2526 | 64 | |
AK029916.1 | - | 4 - 5 | 2507 | 64 | ||
AK049035.1 | - | 4 - 5 | 2577 | 64 | ||
AK134840.1 | - | 4 - 5 | 2515 | 64 | ||
BC138063.1 | - | 4 - 5 | 2480 | 8861082 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |