Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172466.2 | NP_766054.2 | 17 - 18 | 3038 | 73 | |
GenBank mRNA | AK036137.1 | - | 14 - 15 | 2117 | 73 | |
AK053851.1 | - | 16 - 17 | 3150 | 73 | ||
AK140876.1 | - | 17 - 18 | 2814 | 73 | ||
AK163023.1 | - | 2 - 3 | 266 | 73 | ||
BC094674.1 | - | 17 - 18 | 2693 | 30548647 | 73 | |
BC144816.1 | - | 15 - 16 | 2296 | 73 | ||
BC144817.1 | - | 14 - 15 | 2168 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |