Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105196.1 | NP_001098666.1 | 6 - 7 | 1240 | 80 | |
NM_001105197.1 | NP_001098667.1 | 7 - 8 | 1031 | 80 | ||
NM_001271489.1 | NP_001258418.1 | 7 - 8 | 1326 | 80 | ||
NM_001271490.1 | NP_001258419.1 | 7 - 8 | 1498 | 80 | ||
NM_001271491.1 | NP_001258420.1 | 6 - 7 | 960 | 80 | ||
NM_172472.3 | NP_766060.2 | 7 - 8 | 1345 | 80 | ||
GenBank mRNA | AK089762.1 | - | 5 - 6 | 1002 | 80 | |
AK139756.1 | - | 6 - 7 | 1237 | 80 | ||
AK144657.1 | - | 7 - 8 | 1310 | 80 | ||
AK144751.1 | - | 7 - 8 | 1313 | 80 | ||
AK145029.1 | - | 7 - 8 | 1466 | 80 | ||
AK145832.1 | - | 7 - 8 | 1310 | 80 | ||
AK147405.1 | - | 5 - 6 | 3026 | 80 | ||
AK150372.1 | - | 7 - 8 | 1097 | 80 | ||
AK151447.1 | - | 7 - 8 | 1016 | 80 | ||
AK152221.1 | - | 7 - 8 | 1016 | 80 | ||
AK153051.1 | - | 7 - 8 | 1097 | 80 | ||
AK154690.1 | - | 6 - 7 | 1896 | 80 | ||
AK155883.1 | - | 6 - 7 | 962 | 80 | ||
AK159162.1 | - | 7 - 8 | 1226 | 80 | ||
BC056358.1 | - | 7 - 8 | 1333 | 6813486 | 80 | |
BC063047.1 | - | 7 - 8 | 1333 | 6813486 | 80 | |
S76673.1 | - | 5 - 6 | 683 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |