Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282994.1 | NP_001269923.1 | 5 - 6 | 855 | 114 | |
NM_172496.3 | NP_766084.3 | 5 - 6 | 1024 | 114 | ||
GenBank mRNA | AK028833.1 | - | 5 - 6 | 871 | 114 | |
AK049730.1 | - | 5 - 6 | 817 | 114 | ||
AK122334.1 | - | 5 - 6 | 855 | 114 | ||
AY308745.1 | - | 5 - 6 | 864 | 114 | ||
BC145118.1 | - | 5 - 6 | 864 | 9053666 | 114 | |
BC150706.1 | - | 5 - 6 | 864 | 9087617 | 114 | |
CF534605.1 | - | 1 - 2 | 102 | 114 | ||
U26967.1 | - | 4 - 5 | 597 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |