Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172506.2 | NP_766094.1 | 14 - 15 | 2951 | 95 | |
GenBank mRNA | AF388037.1 | - | 13 - 14 | 2404 | 95 | |
AK028770.1 | - | 14 - 15 | 2972 | 95 | ||
AK028889.1 | - | 14 - 15 | 2952 | 95 | ||
AK131182.1 | - | 9 - 10 | 5171 | 95 | ||
BC026443.1 | - | 3 - 4 | 352 | 95 | ||
BC056138.1 | - | 5 - 6 | 854 | 95 | ||
BC062892.1 | - | 14 - 15 | 2931 | 6418202 | 95 | |
BC078631.1 | - | 14 - 15 | 2895 | 6515418 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |