Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172523.3 | NP_766111.1 | 8 - 9 | 1003 | 116 | |
GenBank mRNA | AJ555564.1 | - | 7 - 8 | 867 | 116 | |
AK033871.1 | - | 8 - 9 | 959 | 116 | ||
AK035644.1 | - | 8 - 9 | 986 | 116 | ||
AK041274.1 | - | 8 - 9 | 955 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Mus musculus Probe spans exons |