Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167994.1 | NP_001161466.1 | 12 - 13 | 1491 | 110 | |
NM_172540.2 | NP_766128.2 | 11 - 12 | 1415 | 110 | ||
GenBank mRNA | AK028999.1 | - | 11 - 12 | 1371 | 110 | |
AK049523.1 | - | 11 - 12 | 1413 | 110 | ||
AK129030.1 | - | 11 - 12 | 1608 | 110 | ||
BC030387.1 | - | 10 - 11 | 1076 | 110 | ||
BC056937.1 | - | 12 - 13 | 1432 | 5716144 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |