Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172557.2 | NP_766145.1 | 16 - 17 | 1924 | 98 | |
GenBank mRNA | AJ250024.1 | - | 15 - 16 | 1600 | 98 | |
AK046633.1 | - | 16 - 17 | 1925 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |