Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172562.3 | NP_766150.1 | 2 - 3 | 140 | 63 | |
GenBank mRNA | AK142262.1 | - | 2 - 3 | 172 | 63 | |
AK157938.1 | - | 2 - 3 | 133 | 63 | ||
BC038821.1 | - | 2 - 3 | 143 | 3990023 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |