Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172564.3 | NP_766152.2 | 1 - 2 | 69 | 76 | |
GenBank mRNA | AK036834.1 | - | 1 - 2 | 66 | 76 | |
BC055820.1 | - | 1 - 2 | 51 | 4209161 | 76 | |
BY133848.1 | - | 1 - 2 | 69 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |