Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172582.3 | NP_766170.2 | 8 - 9 | 999 | 64 | |
GenBank mRNA | AK077863.1 | - | 8 - 9 | 923 | 64 | |
AK131889.1 | - | 8 - 9 | 961 | 64 | ||
AK167380.1 | - | 8 - 9 | 927 | 64 | ||
AK170311.1 | - | 7 - 8 | 1464 | 64 | ||
BC024135.1 | - | 8 - 9 | 900 | 64 | ||
BC051055.1 | - | 8 - 9 | 915 | 64 | ||
BC057914.1 | - | 5 - 6 | 663 | 64 | ||
BC088736.1 | - | 4 - 5 | 2248 | 64 | ||
BC106183.1 | - | 8 - 9 | 983 | 64 | ||
BC132168.1 | - | 8 - 9 | 999 | 40130445 | 64 | |
BC132170.1 | - | 8 - 9 | 999 | 40130447 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |