Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253755.1 | NP_001240684.1 | 7 - 8 | 935 | 69 | |
NM_172605.3 | NP_766193.3 | 7 - 8 | 935 | 69 | ||
GenBank mRNA | AK032892.1 | - | 7 - 8 | 880 | 69 | |
AK033390.1 | - | 7 - 8 | 880 | 69 | ||
BC005670.1 | - | 7 - 8 | 836 | 3153274 | 69 | |
BC066144.1 | - | 7 - 8 | 883 | 30435963 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |