Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330027C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172616.2 | NP_766204.2 | 1 - 2 | 193 | 71 | |
GenBank mRNA | AK049356.1 | - | 1 - 2 | 195 | 71 | |
AK160639.1 | - | 1 - 2 | 179 | 71 | ||
BC137719.1 | - | 1 - 2 | 202 | 8860738 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |