Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172662.3 | NP_766250.2 | 5 - 6 | 715 | 102 | |
GenBank mRNA | AK028468.1 | - | 5 - 6 | 638 | 102 | |
AK054272.1 | - | 5 - 6 | 716 | 102 | ||
BC052069.1 | - | 2 - 3 | 280 | 5715781 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |