Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172663.4 | NP_766251.3 | 8 - 9 | 1555 | 87 | |
GenBank mRNA | AK031025.1 | - | 8 - 9 | 1528 | 87 | |
AK047947.1 | - | 8 - 9 | 1223 | 87 | ||
AK141073.1 | - | 846 | 87 | |||
AK164886.1 | - | 4 - 5 | 582 | 87 | ||
BC048785.1 | - | 7 - 8 | 994 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |