Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172687.1 | NP_766275.1 | 3 - 4 | 398 | 88 | |
GenBank mRNA | AK028680.1 | - | 3 - 4 | 398 | 88 | |
AK035318.1 | - | 3 - 4 | 393 | 88 | ||
AK142287.1 | - | 3 - 4 | 336 | 88 | ||
AK166475.1 | - | 3 - 4 | 353 | 88 | ||
AK170488.1 | - | 3 - 4 | 393 | 88 | ||
BC060960.1 | - | 3 - 4 | 400 | 30312962 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |