Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009316.1 | NP_033342.1 | 1 - 2 | 278 | 100 | |
NM_172688.3 | NP_766276.1 | 1 - 2 | 278 | 100 | ||
GenBank mRNA | AK053911.1 | - | 1 - 2 | 264 | 100 | |
AK137046.1 | - | 1 - 2 | 279 | 100 | ||
AK145302.1 | - | 1 - 2 | 289 | 100 | ||
AK159277.1 | - | 1 - 2 | 242 | 100 | ||
BC006665.1 | - | 1 - 2 | 269 | 3499247 | 100 | |
D76446.1 | - | 1 - 2 | 278 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |