Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172692.3 | NP_766280.2 | 15 - 16 | 2795 | 81 | |
GenBank mRNA | AK089192.1 | - | 15 - 16 | 2481 | 81 | |
AK158174.1 | - | 13 - 14 | 3166 | 81 | ||
AK173213.1 | - | 15 - 16 | 2781 | 81 | ||
AK188452.1 | - | 2 - 3 | 242 | 81 | ||
BC031397.1 | - | 13 - 14 | 2798 | 81 | ||
BC034105.1 | - | 12 - 13 | 2546 | 81 | ||
BC056935.1 | - | 15 - 16 | 2318 | 5703478 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |