Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172700.2 | NP_766288.1 | 5 - 6 | 870 | 149 | |
GenBank mRNA | AJ487544.1 | - | 5 - 6 | 836 | 149 | |
AK083566.1 | - | 5 - 6 | 870 | 149 | ||
AK170946.1 | - | 5 - 6 | 829 | 149 | ||
AY029194.1 | - | 5 - 6 | 626 | 149 | ||
BC023740.1 | - | 5 - 6 | 813 | 149 | ||
BC138578.1 | - | 5 - 6 | 738 | 8861598 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |