Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289577.1 | NP_001276506.1 | 7 - 8 | 895 | 83 | |
NM_001289578.1 | NP_001276507.1 | 6 - 7 | 679 | 83 | ||
NM_001289579.1 | NP_001276508.1 | 8 - 9 | 968 | 83 | ||
NM_001289580.1 | NP_001276509.1 | 8 - 9 | 1065 | 83 | ||
NM_172717.4 | NP_766305.2 | 7 - 8 | 895 | 83 | ||
GenBank mRNA | AK048930.1 | - | 8 - 9 | 1046 | 84 | |
AK077629.1 | - | 7 - 8 | 867 | 83 | ||
AK133154.1 | - | 8 - 9 | 950 | 83 | ||
AK147892.1 | - | 6 - 7 | 654 | 83 | ||
AK154038.1 | - | 7 - 8 | 877 | 83 | ||
AK155295.1 | - | 7 - 8 | 867 | 83 | ||
AK155525.1 | - | 7 - 8 | 867 | 83 | ||
AK169182.1 | - | 8 - 9 | 921 | 83 | ||
AK169193.1 | - | 7 - 8 | 850 | 83 | ||
BC049792.1 | - | 7 - 8 | 849 | 6510480 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |