Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172723.4 | NP_766311.2 | 9 - 10 | 971 | 91 | |
GenBank mRNA | AK076754.1 | - | 8 - 9 | 957 | 91 | |
AK076768.1 | - | 9 - 10 | 728 | 91 | ||
BC056934.1 | - | 8 - 9 | 697 | 91 | ||
BC079634.1 | - | 8 - 9 | 765 | 91 | ||
BC144874.1 | - | 8 - 9 | 713 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |