Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172778.2 | NP_766366.2 | 9 - 10 | 1112 | 68 | |
GenBank mRNA | AK031833.1 | - | 9 - 10 | 1096 | 68 | |
AK054050.1 | - | 9 - 10 | 1111 | 68 | ||
BC113182.1 | - | 9 - 10 | 1101 | 40061991 | 68 | |
BC113788.1 | - | 9 - 10 | 1101 | 40061990 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |