Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172802.4 | NP_766390.2 | 3 - 4 | 1281 | 66 | |
GenBank mRNA | AK044556.1 | - | 4 - 5 | 1212 | 66 | |
AK050002.1 | - | 3 - 4 | 1282 | 66 | ||
BB279997.2 | - | 2 - 3 | 245 | 66 | ||
BC109356.1 | - | 3 - 4 | 1242 | 40057445 | 66 | |
BC109357.1 | - | 3 - 4 | 1242 | 40057447 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |