Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172824.3 | NP_766412.2 | 3 - 4 | 341 | 60 | |
GenBank mRNA | AK029725.1 | - | 3 - 4 | 342 | 60 | |
AK090294.1 | - | 3 - 4 | 248 | 60 | ||
BB615458.1 | - | 3 - 4 | 342 | 60 | ||
BB618371.1 | - | 3 - 4 | 243 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |