Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172840.2 | NP_766428.2 | 4 - 5 | 436 | 68 | |
GenBank mRNA | AJ536329.1 | - | 4 - 5 | 436 | 68 | |
AK029297.1 | - | 4 - 5 | 383 | 68 | ||
AK133486.1 | - | 4 - 5 | 343 | 68 | ||
AK145058.1 | - | 4 - 5 | 606 | 68 | ||
AK160971.1 | - | 4 - 5 | 441 | 68 | ||
AK166339.1 | - | 4 - 5 | 514 | 68 | ||
BC115868.1 | - | 4 - 5 | 385 | 40081354 | 68 | |
BC116636.1 | - | 4 - 5 | 386 | 40081359 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |