Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172844.2 | NP_766432.1 | 2 - 3 | 204 | 103 | |
GenBank mRNA | AK029219.1 | - | 2 - 3 | 205 | 103 | |
AK161224.1 | - | 2 - 3 | 204 | 103 | ||
BC112405.1 | - | 2 - 3 | 194 | 40060428 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |