Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199224.1 | NP_001186153.1 | 12 - 13 | 1749 | 86 | |
GenBank mRNA | AK052433.1 | - | 6 - 7 | 1235 | 86 | |
DQ867033.1 | - | 6 - 7 | 1235 | 86 | ||
FN429927.1 | - | 12 - 13 | 1749 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |