Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172926.3 | NP_766514.2 | 13 - 14 | 1412 | 76 | |
GenBank mRNA | AK046839.1 | - | 13 - 14 | 1354 | 76 | |
AK049371.1 | - | 13 - 14 | 1391 | 76 | ||
AK082389.1 | - | 12 - 13 | 1306 | 76 | ||
BC080799.1 | - | 13 - 14 | 1397 | 4948538 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |