Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200014364.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014364.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014364.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014370.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014370.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014370.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014371.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014371.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014371.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014366.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014366.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014366.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014367.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014367.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014367.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014368.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014368.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014368.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014372.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014372.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014372.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014374.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014374.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014374.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014378.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014378.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014378.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014369.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014369.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014369.mm.1 |
TC0200001849.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014376.mm.1 |
NM_172980.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014376.mm.1 |
NM_172980.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014376.mm.1 |
TC0200001849.mm.1 |
MTA TC |