Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172988.4 | NP_766576.1 | 3 - 4 | 832 | 88 | |
NR_104378.1 | 3 - 4 | 832 | 88 | |||
NR_104379.1 | 3 - 4 | 832 | 88 | |||
NR_104380.1 | 3 - 4 | 832 | 88 | |||
GenBank mRNA | AK028786.1 | - | 3 - 4 | 816 | 88 | |
AK032391.1 | - | 3 - 4 | 811 | 88 | ||
AK051429.1 | - | 3 - 4 | 783 | 88 | ||
AK083661.1 | - | 3 - 4 | 806 | 88 | ||
BC033265.1 | - | 3 - 4 | 759 | 3983942 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |