Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172988.4 | NP_766576.1 | 6 - 7 | 1630 | 86 | |
NR_104378.1 | 6 - 7 | 1630 | 86 | |||
NR_104379.1 | 6 - 7 | 1630 | 86 | |||
NR_104380.1 | 6 - 7 | 1593 | 86 | |||
GenBank mRNA | AK028786.1 | - | 6 - 7 | 1614 | 86 | |
AK032391.1 | - | 6 - 7 | 1609 | 86 | ||
AK051429.1 | - | 6 - 7 | 1581 | 86 | ||
AK083661.1 | - | 6 - 7 | 1567 | 86 | ||
BC033265.1 | - | 6 - 7 | 1557 | 3983942 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |