Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172998.3 | NP_766586.2 | 4 - 5 | 328 | 90 | |
GenBank mRNA | AK046850.1 | - | 4 - 5 | 329 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |