Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173400.2 | NP_775576.2 | 10 - 11 | 1403 | 73 | |
GenBank mRNA | AK036635.1 | - | 1 - 2 | 68 | 73 | |
AK051016.1 | - | 10 - 11 | 1424 | 73 | ||
AK077681.1 | - | 10 - 11 | 1425 | 73 | ||
AK136163.1 | - | 1 - 2 | 123 | 73 | ||
AK169222.1 | - | 10 - 11 | 1405 | 73 | ||
AK173206.1 | - | 10 - 11 | 1375 | 73 | ||
BC064725.1 | - | 5 - 6 | 600 | 73 | ||
BC068237.1 | - | 10 - 11 | 1403 | 30550558 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |