Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163512.1 | NP_001156984.1 | 12 - 13 | 1545 | 103 | |
NM_173402.2 | NP_775578.2 | 15 - 16 | 3592 | 103 | ||
GenBank mRNA | AK004813.2 | - | 13 - 14 | 1498 | 103 | |
AK014569.1 | - | 12 - 13 | 1547 | 103 | ||
AK145890.1 | - | 15 - 16 | 3541 | 103 | ||
BC040396.1 | - | 15 - 16 | 4461061 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |