Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500015627.mm.1 |
TC0500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015627.mm.1 |
NM_173405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015627.mm.1 |
NM_173405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015629.mm.1 |
TC0500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015629.mm.1 |
NM_173405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015629.mm.1 |
NM_173405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015628.mm.1 |
TC0500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015628.mm.1 |
NM_173405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015628.mm.1 |
NM_173405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015620.mm.1 |
TC0500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015620.mm.1 |
NM_173405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015620.mm.1 |
NM_173405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015621.mm.1 |
TC0500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015621.mm.1 |
NM_173405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015621.mm.1 |
NM_173405.2 |
Refseq |