Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173740.3 | NP_776101.3 | 4 - 5 | 559 | 80 | |
GenBank mRNA | AK004337.1 | - | 4 - 5 | 421 | 80 | |
AK146612.1 | - | 4 - 5 | 560 | 80 | ||
AK155052.1 | - | 4 - 5 | 422 | 80 | ||
AK164457.1 | - | 4 - 5 | 396 | 80 | ||
BC029100.1 | - | 4 - 5 | 429 | 3481702 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |