Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100008035.mm.1 |
NM_174874.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008035.mm.1 |
NM_174874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008035.mm.1 |
TC0100000944.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008033.mm.1 |
NM_174874.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008033.mm.1 |
NM_174874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008033.mm.1 |
TC0100000944.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008034.mm.1 |
NM_174874.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008034.mm.1 |
NM_174874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008034.mm.1 |
TC0100000944.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008036.mm.1 |
NM_174874.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008036.mm.1 |
NM_174874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008036.mm.1 |
TC0100000944.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008037.mm.1 |
NM_174874.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008037.mm.1 |
NM_174874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008037.mm.1 |
TC0100000944.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008040.mm.1 |
NM_174874.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008040.mm.1 |
NM_174874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008040.mm.1 |
TC0100000944.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008041.mm.1 |
NM_174874.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008041.mm.1 |
NM_174874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008041.mm.1 |
TC0100000944.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008030.mm.1 |
TC0100000944.mm.1 |
MTA TC |