Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033329.3 | NP_001028501.1 | 3 - 4 | 669 | 62 | |
NM_001290384.1 | NP_001277313.1 | 2 - 3 | 291 | 62 | ||
NM_001290385.1 | NP_001277314.1 | 2 - 3 | 1115 | 62 | ||
GenBank mRNA | AK038840.1 | - | 3 - 4 | 587 | 62 | |
AK122280.1 | - | 2 - 3 | 226 | 62 | ||
AK139416.1 | - | 3 - 4 | 617 | 62 | ||
AK163874.1 | - | 2 - 3 | 272 | 62 | ||
BC067031.1 | - | 2 - 3 | 937 | 62 | ||
BC141385.1 | - | 2 - 3 | 311 | 9055998 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |