Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173774.3 | NP_776135.2 | 2 - 3 | 572 | 69 | |
GenBank mRNA | AK082651.1 | - | 564 | 69 | ||
AK134677.1 | - | 2 - 3 | 496 | 69 | ||
AK134683.1 | - | 2 - 3 | 562 | 69 | ||
AK158002.1 | - | 2 - 3 | 562 | 69 | ||
BC051188.1 | - | 2 - 3 | 532 | 69 | ||
CX241103.1 | - | 2 - 3 | 490 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |